The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289080.1 | NP_001276009.1 | 16 - 17 | 2638 | 74 |
NM_001289081.1 | NP_001276010.1 | 15 - 16 | 2501 | 74 | |
NM_014461.3 | NP_055276.1 | 16 - 17 | 2647 | 74 | |
XM_005265058.3 | XP_005265115.1 | 17 - 18 | 2770 | 74 | |
XM_011533590.2 | XP_011531892.1 | 16 - 17 | 2294 | 74 | |
XM_011533591.2 | XP_011531893.1 | 15 - 16 | 2609 | 74 | |
XM_017006171.1 | XP_016861660.1 | 21 - 22 | 3197 | 74 | |
XM_017006172.1 | XP_016861661.1 | 16 - 17 | 2772 | 74 | |
XM_017006173.1 | XP_016861662.1 | 15 - 16 | 2278 | 74 | |
XM_017006174.1 | XP_016861663.1 | 15 - 16 | 2261 | 74 | |
XM_017006175.1 | XP_016861664.1 | 13 - 14 | 1935 | 74 | |
XM_017006176.1 | XP_016861665.1 | 14 - 15 | 2079 | 74 | |
GenBank mRNA | AB003592.1 | - | 16 - 17 | 2360 | 74 |
AK294786.1 | - | 15 - 16 | 2501 | 74 | |
AK314220.1 | - | 16 - 17 | 2361 | 74 | |
BC113118.1 | - | 16 - 17 | 2273 | 74 |