Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTN6
Entrez Gene ID
27255
Gene Name
contactin 6
Gene Aliases
NB3
Chromosome Location
Chr.3: 978021 - 1403610 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289080.1 NP_001276009.1 19 - 20 3060 110
NM_001289081.1 NP_001276010.1 18 - 19 2923 110
NM_014461.3 NP_055276.1 19 - 20 3069 110
XM_005265058.3 XP_005265115.1 20 - 21 3192 110
XM_011533590.2 XP_011531892.1 19 - 20 2716 110
XM_011533591.2 XP_011531893.1 18 - 19 3031 110
XM_017006171.1 XP_016861660.1 24 - 25 3619 110
XM_017006172.1 XP_016861661.1 19 - 20 3194 110
XM_017006173.1 XP_016861662.1 18 - 19 2700 110
XM_017006174.1 XP_016861663.1 18 - 19 2683 110
XM_017006175.1 XP_016861664.1 16 - 17 2357 110
XM_017006176.1 XP_016861665.1 17 - 18 2501 110
GenBank mRNA AB003592.1 - 19 - 20 2782 110
AK294786.1 - 18 - 19 2923 110
AK314220.1 - 19 - 20 2783 110
BC113118.1 - 19 - 20 2695 110

View Full Product Details