The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289080.1 | NP_001276009.1 | 7 - 8 | 1310 | 87 |
NM_001289081.1 | NP_001276010.1 | 6 - 7 | 1173 | 87 | |
NM_014461.3 | NP_055276.1 | 7 - 8 | 1319 | 87 | |
XM_005265058.3 | XP_005265115.1 | 8 - 9 | 1442 | 87 | |
XM_011533590.2 | XP_011531892.1 | 7 - 8 | 966 | 87 | |
XM_011533591.2 | XP_011531893.1 | 6 - 7 | 1281 | 87 | |
XM_017006171.1 | XP_016861660.1 | 12 - 13 | 1869 | 87 | |
XM_017006172.1 | XP_016861661.1 | 7 - 8 | 1444 | 87 | |
XM_017006174.1 | XP_016861663.1 | 6 - 7 | 933 | 87 | |
XM_017006177.1 | XP_016861666.1 | 7 - 8 | 1058 | 87 | |
GenBank mRNA | AB003592.1 | - | 7 - 8 | 1032 | 87 |
AK294786.1 | - | 6 - 7 | 1173 | 87 | |
AK314220.1 | - | 7 - 8 | 1033 | 87 | |
BC113118.1 | - | 7 - 8 | 945 | 87 |