The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286752.1 | NP_001273681.1 | 9 - 10 | 1630 | 153 |
NM_017438.4 | NP_059134.1 | 8 - 9 | 1487 | 153 | |
NR_040087.2 | 7 - 8 | 1398 | 153 | ||
XM_011529636.1 | XP_011527938.1 | 8 - 9 | 1536 | 153 | |
XM_011529637.1 | XP_011527939.1 | 8 - 9 | 1601 | 153 | |
XM_011529638.2 | XP_011527940.1 | 8 - 9 | 1509 | 153 | |
XM_011529639.1 | XP_011527941.1 | 8 - 9 | 1287 | 153 | |
XM_011529640.2 | XP_011527942.1 | 8 - 9 | 1552 | 153 | |
XM_011529642.1 | XP_011527944.1 | 9 - 10 | 1630 | 153 | |
XM_011529643.1 | XP_011527945.1 | 9 - 10 | 1581 | 153 | |
XM_011529644.1 | XP_011527946.1 | 9 - 10 | 1142 | 153 | |
XM_017028403.1 | XP_016883892.1 | 8 - 9 | 1078 | 153 | |
XM_017028404.1 | XP_016883893.1 | 9 - 10 | 1186 | 153 | |
GenBank mRNA | AF391112.1 | - | 7 - 8 | 965 | 153 |
AK001660.1 | - | 8 - 9 | 1054 | 153 | |
AK095215.1 | - | 7 - 8 | 2354 | 153 | |
BC036556.2 | - | 9 - 10 | 1304 | 153 |