The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005861.2 | NP_001005861.1 | 2 - 3 | 438 | 74 |
NM_002958.3 | NP_002949.2 | 2 - 3 | 438 | 74 | |
GenBank mRNA | CN309980.1 | - | 2 - 3 | 282 | 74 |
S59184.1 | - | 2 - 3 | 471 | 74 | |
X69970.1 | - | 2 - 3 | 438 | 74 | |
X96588.1 | - | 3 - 4 | 343 | 74 |