Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RYK
Entrez Gene ID
6259
Gene Name
receptor-like tyrosine kinase
Gene Aliases
D3S3195, JTK5, JTK5A, RYK1
Chromosome Location
Chr.3: 134157133 - 134250834 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005861.2 NP_001005861.1 6 - 7 876 155
NM_002958.3 NP_002949.2 6 - 7 876 155
GenBank mRNA BC021700.2 - 1 - 2 142 155
S59184.1 - 6 - 7 909 155
X69970.1 - 6 - 7 876 155
X96588.1 - 7 - 8 781 155

View Full Product Details