The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182575.2 | NP_872381.2 | 1 - 2 | 92 | |
GenBank mRNA | AB195682.1 | - | 1 - 2 | 338 | 92 |
BC034769.1 | - | 1 - 2 | 480 | 92 |