The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080927.3 | NP_563615.3 | 10 - 11 | 1730 | 65 |
XM_011512419.2 | XP_011510721.1 | 9 - 10 | 1446 | 65 | |
GenBank mRNA | AB073146.1 | - | 10 - 11 | 1730 | 65 |
AF387547.1 | - | 10 - 11 | 1378 | 65 | |
AK056350.1 | - | 10 - 11 | 1723 | 65 | |
AK302649.1 | - | 10 - 11 | 1210 | 65 | |
BC029658.1 | - | 10 - 11 | 1249 | 65 | |
BX647140.1 | - | 1 - 2 | 3963 | 65 | |
D29810.1 | - | 9 - 10 | 1170 | 65 |