Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCBLD2
Entrez Gene ID
131566
Gene Name
discoidin, CUB and LCCL domain containing 2
Gene Aliases
CLCP1, ESDN
Chromosome Location
Chr.3: 98795970 - 98901689 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080927.3 NP_563615.3 14 - 15 2084 94
XM_011512419.2 XP_011510721.1 13 - 14 1800 94
GenBank mRNA AB073146.1 - 14 - 15 2084 94
AF387547.1 - 14 - 15 1732 94
AK302649.1 - 14 - 15 1564 94

View Full Product Details