The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003009.2 | NP_003000.1 | 5 - 6 | 482 | 82 |
GenBank mRNA | AF015283.1 | - | 5 - 6 | 331 | 82 |
AK026717.1 | - | 4 - 5 | 2044 | 82 | |
BC000581.2 | - | 5 - 6 | 347 | 82 | |
BC032546.1 | - | 5 - 6 | 372 | 82 | |
BC039597.1 | - | 5 - 6 | 350 | 82 | |
BC047893.1 | - | 5 - 6 | 349 | 82 | |
BU185196.1 | - | - | 82 | ||
U67171.1 | - | 5 - 6 | 342 | 82 |