The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001280794.2 | NP_001267723.2 | 9 - 10 | 2324 | 59 |
NM_001280795.2 | NP_001267724.2 | 6 - 7 | 1696 | 59 | |
NM_004445.5 | NP_004436.4 | 7 - 8 | 1765 | 59 | |
XM_006715881.2 | XP_006715944.1 | 10 - 11 | 2760 | 59 | |
XM_011515879.1 | XP_011514181.1 | 11 - 12 | 2799 | 59 | |
XM_011515880.1 | XP_011514182.1 | 10 - 11 | 2777 | 59 | |
XM_011515881.2 | XP_011514183.1 | 10 - 11 | 2761 | 59 | |
GenBank mRNA | BC056159.1 | - | 9 - 10 | 2193 | 59 |
BC110606.1 | - | 6 - 7 | 1690 | 59 | |
BC110607.1 | - | 7 - 8 | 1759 | 59 | |
D83492.1 | - | 10 - 11 | 2336 | 59 | |
EU054308.1 | - | 5 - 6 | 1702 | 59 |