The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305155.1 | NP_001292084.1 | 5 - 6 | 1023 | 112 |
NM_001305156.1 | NP_001292085.1 | 6 - 7 | 904 | 112 | |
NM_017917.3 | NP_060387.2 | 6 - 7 | 1151 | 112 | |
NR_130972.1 | 6 - 7 | 904 | 112 | ||
XM_005267782.3 | XP_005267839.1 | 7 - 8 | 1032 | 112 | |
XM_017021388.1 | XP_016876877.1 | 7 - 8 | 1032 | 112 | |
XM_017021389.1 | XP_016876878.1 | 6 - 7 | 1715 | 112 | |
GenBank mRNA | AK000651.1 | - | 6 - 7 | 844 | 112 |
AK225512.1 | - | 6 - 7 | 844 | 112 | |
AK293717.1 | - | 6 - 7 | 808 | 112 | |
AY157304.1 | - | 6 - 7 | 922 | 112 | |
AY518535.1 | - | 6 - 7 | 561 | 112 | |
AY518536.1 | - | 5 - 6 | 460 | 112 | |
AY518537.1 | - | 6 - 7 | 555 | 112 | |
BC006823.1 | - | 6 - 7 | 602 | 112 | |
BC010293.2 | - | 6 - 7 | 589 | 112 | |
BC012563.1 | - | 6 - 7 | 583 | 112 | |
BC063438.1 | - | 5 - 6 | 734 | 112 | |
BC100021.1 | - | 6 - 7 | 622 | 112 |