The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000255.3 | NP_000246.2 | 12 - 13 | 2391 | 99 |
XM_005249143.3 | XP_005249200.1 | 12 - 13 | 2296 | 99 | |
GenBank mRNA | AB208845.1 | - | 6 - 7 | 4738 | 99 |
AK292568.1 | - | 12 - 13 | 2367 | 99 | |
AK312611.1 | - | 12 - 13 | 2366 | 99 | |
BC016282.1 | - | 12 - 13 | 2254 | 99 | |
BT007434.1 | - | 11 - 12 | 2126 | 99 | |
BX647789.1 | - | 11 - 12 | 3702 | 99 | |
CA444307.1 | - | 1 - 2 | 98 | ||
KU178201.1 | - | 11 - 12 | 2126 | 99 | |
KU178202.1 | - | 12 - 13 | 2202 | 99 | |
M65131.1 | - | 12 - 13 | 2202 | 99 |