The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159643.1 | NP_001153115.1 | 14 - 15 | 1955 | 172 |
NM_001159644.1 | NP_001153116.1 | 7 - 8 | 1022 | 172 | |
NM_018349.3 | NP_060819.3 | 14 - 15 | 1955 | 172 | |
XM_005254955.3 | XP_005255012.1 | 15 - 16 | 2436 | 172 | |
XM_005254960.2 | XP_005255017.1 | 7 - 8 | 1027 | 172 | |
XM_006720603.2 | XP_006720666.1 | 16 - 17 | 2192 | 172 | |
XM_011521770.1 | XP_011520072.1 | 14 - 15 | 1955 | 172 | |
XM_011521771.2 | XP_011520073.1 | 16 - 17 | 2361 | 172 | |
XM_011521772.2 | XP_011520074.1 | 15 - 16 | 2251 | 172 | |
XM_011521773.2 | XP_011520075.1 | 14 - 15 | 1955 | 172 | |
XM_011521774.2 | XP_011520076.1 | 14 - 15 | 1955 | 172 | |
XM_011521775.2 | XP_011520077.1 | 8 - 9 | 1707 | 172 | |
XM_017022403.1 | XP_016877892.1 | 13 - 14 | 1919 | 172 | |
XM_017022404.1 | XP_016877893.1 | 16 - 17 | 2361 | 172 | |
XM_017022405.1 | XP_016877894.1 | 15 - 16 | 2366 | 172 | |
GenBank mRNA | AK002037.1 | - | 7 - 8 | 730 | 172 |
AY656717.1 | - | 14 - 15 | 1890 | 172 | |
BC025708.1 | - | 7 - 8 | 983 | 172 | |
BC041387.1 | - | 16 - 17 | 2378 | 172 | |
BC111024.1 | - | 7 - 8 | 855 | 172 | |
BC131527.1 | - | 14 - 15 | 1890 | 172 |