The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004163.4 | NP_004154.2 | 4 - 5 | 588 | 110 |
XM_005266733.1 | XP_005266790.1 | 5 - 6 | 509 | 110 | |
XM_006722518.1 | XP_006722581.1 | 4 - 5 | 866 | 110 | |
XM_011526131.1 | XP_011524433.1 | 4 - 5 | 866 | 110 | |
XM_017025913.1 | XP_016881402.1 | 7 - 8 | 1342 | 110 | |
XM_017025914.1 | XP_016881403.1 | 6 - 7 | 737 | 110 | |
GenBank mRNA | AF329499.1 | - | 4 - 5 | 588 | 110 |
AF498954.1 | - | 3 - 4 | 345 | 110 | |
AK314115.1 | - | 4 - 5 | 866 | 110 | |
BC027474.1 | - | 4 - 5 | 456 | 110 | |
U57093.2 | - | 4 - 5 | 437 | 110 |