The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039.3 | NP_001030.2 | 10 - 11 | 1571 | 84 |
GenBank mRNA | BC059391.1 | - | 10 - 11 | 1571 | 84 |
BC069652.1 | - | 9 - 10 | 1432 | 84 | |
DQ402524.1 | - | 9 - 10 | 1432 | 84 | |
L36592.1 | - | 9 - 10 | 1454 | 84 | |
X87160.1 | - | 9 - 10 | 1432 | 84 |