Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB3GAP2
Entrez Gene ID
25782
Gene Name
RAB3 GTPase activating non-catalytic protein subunit 2
Gene Aliases
RAB3-GAP150, RAB3GAP150, SPG69, WARBM2, p150
Chromosome Location
Chr.1: 220148268 - 220272501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012414.3 NP_036546.2 22 - 23 2586 93
GenBank mRNA AB020646.2 - 22 - 23 2422 93
AF004828.1 - 22 - 23 2464 93
AF255648.1 - 22 - 23 2495 93
AK027485.1 - 4 - 5 508 93
AK291234.1 - 22 - 23 2537 93
BC131573.1 - 22 - 23 2469 93
BC146760.1 - 22 - 23 2422 93

View Full Product Details