The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012414.3 | NP_036546.2 | 29 - 30 | 3421 | 140 |
GenBank mRNA | AB020646.2 | - | 29 - 30 | 3257 | 140 |
AF004828.1 | - | 29 - 30 | 3299 | 140 | |
AF255648.1 | - | 29 - 30 | 3330 | 140 | |
AK027485.1 | - | 11 - 12 | 1343 | 140 | |
AK291234.1 | - | 29 - 30 | 3372 | 140 | |
AL117631.1 | - | 6 - 7 | 470 | 140 | |
BC131573.1 | - | 29 - 30 | 3300 | 140 | |
BC146760.1 | - | 29 - 30 | 3257 | 140 |