The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040458.1 | NP_001035548.1 | 1 - 2 | 327 | 146 |
NM_016442.3 | NP_057526.3 | 1 - 2 | 327 | 146 | |
XM_005272015.4 | XP_005272072.1 | 1 - 2 | 278 | 146 | |
XM_017009583.1 | XP_016865072.1 | 1 - 2 | 273 | 146 | |
GenBank mRNA | DA704693.1 | - | 1 - 2 | 381 | 146 |