Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB41L4B
Entrez Gene ID
54566
Gene Name
erythrocyte membrane protein band 4.1 like 4B
Gene Aliases
CG1, EHM2
Chromosome Location
Chr.9: 109171974 - 109320964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018424.3 NP_060894.2 3 - 4 979 102
NM_019114.4 NP_061987.3 3 - 4 979 102
XM_011518790.1 XP_011517092.1 3 - 4 476 102
XM_011518791.2 XP_011517093.1 3 - 4 461 102
XM_011518792.2 XP_011517094.1 3 - 4 461 102
XM_011518794.2 XP_011517096.1 3 - 4 461 102
XM_017014813.1 XP_016870302.1 3 - 4 461 102
XM_017014814.1 XP_016870303.1 3 - 4 461 102
XM_017014815.1 XP_016870304.1 3 - 4 461 102
GenBank mRNA AB032179.3 - 3 - 4 979 102
AF153416.1 - 3 - 4 751 102
AF153418.1 - 3 - 4 751 102
AK000388.1 - 3 - 4 363 102

View Full Product Details