The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281517.2 | NP_001268446.1 | 10 - 11 | 1818 | 70 |
NM_001281518.2 | NP_001268447.1 | 4 - 5 | 379 | 70 | |
NM_001319944.1 | NP_001306873.1 | 11 - 12 | 1971 | 70 | |
NM_022778.4 | NP_073615.2 | 11 - 12 | 1971 | 70 | |
GenBank mRNA | AK024038.1 | - | 11 - 12 | 1927 | 70 |
AK025653.1 | - | 5 - 6 | 570 | 70 | |
AK299312.1 | - | 10 - 11 | 1771 | 70 | |
AL133609.1 | - | 7 - 8 | 812 | 70 | |
AL137420.1 | - | 5 - 6 | 569 | 70 | |
BC019902.1 | - | 4 - 5 | 372 | 70 | |
BC064528.1 | - | 11 - 12 | 1922 | 70 | |
DA666742.1 | - | 4 - 5 | 379 | 70 |