Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEP85
Entrez Gene ID
64793
Gene Name
centrosomal protein 85
Gene Aliases
CCDC21
Chromosome Location
Chr.1: 26234153 - 26278810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281517.2 NP_001268446.1 4 - 5 1064 99
NM_001319944.1 NP_001306873.1 5 - 6 1217 99
NM_022778.4 NP_073615.2 5 - 6 1217 99
XM_017002105.1 XP_016857594.1 5 - 6 1217 99
GenBank mRNA AK024038.1 - 5 - 6 1173 99
AK299312.1 - 4 - 5 1017 99
BC064528.1 - 5 - 6 1168 99

View Full Product Details