The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281517.2 | NP_001268446.1 | 8 - 9 | 1679 | 83 |
NM_001281518.2 | NP_001268447.1 | 2 - 3 | 240 | 83 | |
NM_001319944.1 | NP_001306873.1 | 9 - 10 | 1832 | 83 | |
NM_022778.4 | NP_073615.2 | 9 - 10 | 1832 | 83 | |
XM_017002105.1 | XP_016857594.1 | 9 - 10 | 1832 | 83 | |
GenBank mRNA | AK024038.1 | - | 9 - 10 | 1788 | 83 |
AK299312.1 | - | 8 - 9 | 1632 | 83 | |
AL133609.1 | - | 5 - 6 | 673 | 83 | |
AL137420.1 | - | 3 - 4 | 430 | 83 | |
BC019902.1 | - | 2 - 3 | 233 | 83 | |
BC064528.1 | - | 9 - 10 | 1783 | 83 | |
DA666742.1 | - | 2 - 3 | 240 | 83 |