The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184768.1 | NP_001171697.1 | 2 - 3 | 199 | 85 |
NR_033670.1 | 2 - 3 | 199 | 85 | ||
XM_011530976.2 | XP_011529278.1 | 2 - 3 | 174 | 85 | |
GenBank mRNA | AW955663.1 | - | 2 - 3 | 111 | 85 |
DA933518.1 | - | 2 - 3 | 196 | 85 |