The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012463.3 | NP_036595.2 | 9 - 10 | 1286 | 147 |
GenBank mRNA | AF112972.1 | - | 9 - 10 | 1043 | 147 |
AK289391.1 | - | 9 - 10 | 1201 | 147 | |
BC068531.1 | - | 9 - 10 | 1286 | 147 | |
BX328381.1 | - | 3 - 4 | 379 | 147 |