The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001409.3 | NP_001400.3 | 36 - 37 | 4794 | 74 |
XM_006710406.3 | XP_006710469.1 | 33 - 34 | 4429 | 74 | |
XM_011540885.1 | XP_011539187.1 | 37 - 38 | 5099 | 74 | |
XM_011540886.1 | XP_011539188.1 | 36 - 37 | 4970 | 74 | |
XM_011540887.2 | XP_011539189.1 | 32 - 33 | 4307 | 74 | |
XM_017000533.1 | XP_016856022.1 | 36 - 37 | 4967 | 74 | |
GenBank mRNA | AB011539.2 | - | 29 - 30 | 3834 | 74 |
AB231860.1 | - | 36 - 37 | 75 |