Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAP29
Entrez Gene ID
55973
Gene Name
B-cell receptor-associated protein 29
Gene Aliases
BAP29
Chromosome Location
Chr.7: 107579977 - 107623317 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008405.2 NP_001008405.1 3 - 4 364 119
NM_018844.3 NP_061332.2 4 - 5 689 119
NR_027830.1 3 - 4 583 119
XM_006716051.2 XP_006716114.1 4 - 5 685 119
XM_011516406.2 XP_011514708.1 4 - 5 448 119
XM_011516407.1 XP_011514709.1 4 - 5 685 119
XM_011516408.2 XP_011514710.1 4 - 5 685 119
XM_011516409.2 XP_011514711.1 4 - 5 685 119
XM_017012436.1 XP_016867925.1 4 - 5 685 119
XM_017012437.1 XP_016867926.1 4 - 5 685 119
XM_017012438.1 XP_016867927.1 4 - 5 685 119
GenBank mRNA AK289401.1 - 4 - 5 446 119
AK294824.1 - 3 - 4 314 119
AK298594.1 - 1 - 2 90 119
BC008478.1 - 4 - 5 401 119
BM023571.1 - 1 - 2 159 119
BT006981.1 - 3 - 4 350 119
BX647653.1 - 4 - 5 399 118
CK820004.1 - 2 - 3 221 119

View Full Product Details