Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAP29
Entrez Gene ID
55973
Gene Name
B-cell receptor-associated protein 29
Gene Aliases
BAP29
Chromosome Location
Chr.7: 107579977 - 107623317 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018844.3 NP_061332.2 7 - 8 1027 109
NR_027830.1 6 - 7 921 109
GenBank mRNA AF126020.1 - 7 - 8 737 109
AK289401.1 - 7 - 8 784 109
AK294824.1 - 6 - 7 652 109
BC008478.1 - 7 - 8 739 109
BX647653.1 - 7 - 8 737 109

View Full Product Details