Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDEM2
Entrez Gene ID
55741
Gene Name
ER degradation enhancing alpha-mannosidase like protein 2
Gene Aliases
C20orf31, C20orf49, bA4204.1
Chromosome Location
Chr.20: 35115357 - 35147358 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145025.1 NP_001138497.1 6 - 7 828 106
NM_018217.2 NP_060687.2 7 - 8 939 106
NR_026728.1 6 - 7 1155 106
GenBank mRNA AK001645.1 - 7 - 8 921 106
AK023931.1 - 7 - 8 909 106
AK096016.1 - 5 - 6 849 106
AK293583.1 - 6 - 7 810 106
AK298794.1 - 6 - 7 897 106
AK300212.1 - 6 - 7 783 106
AK303816.1 - 5 - 6 581 106
AY007088.1 - 4 - 5 507 106
AY358580.1 - 6 - 7 789 106
BC001371.2 - 7 - 8 906 106
BC016184.1 - 7 - 8 880 106
CR457266.1 - 7 - 8 839 106

View Full Product Details