The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286431.1 | NP_001273360.1 | 20 - 21 | 7759 | 149 |
NM_001286432.1 | NP_001273361.1 | 19 - 20 | 7652 | 149 | |
NM_002912.4 | NP_002903.3 | 18 - 19 | 7524 | 149 | |
XM_011536028.1 | XP_011534330.1 | 17 - 18 | 7618 | 149 | |
XM_011536029.2 | XP_011534331.1 | 17 - 18 | 7615 | 149 | |
XM_011536030.2 | XP_011534332.1 | 17 - 18 | 7618 | 149 | |
XM_011536031.2 | XP_011534333.1 | 18 - 19 | 7746 | 149 | |
XM_011536032.2 | XP_011534334.1 | 18 - 19 | 12336 | 149 | |
XM_017011152.1 | XP_016866641.1 | 18 - 19 | 7743 | 149 | |
XM_017011153.1 | XP_016866642.1 | 18 - 19 | 7286 | 149 | |
XM_017011154.1 | XP_016866643.1 | 18 - 19 | 7171 | 149 | |
GenBank mRNA | AB209256.1 | - | 5 - 6 | 1440 | 149 |
AF035537.1 | - | 16 - 17 | 6861 | 149 | |
AF058701.1 | - | 18 - 19 | 7151 | 149 | |
AF071798.1 | - | 17 - 18 | 7389 | 149 | |
AF078695.1 | - | 19 - 20 | 7654 | 149 | |
AF157476.1 | - | 17 - 18 | 7097 | 149 | |
AF179428.1 | - | 18 - 19 | 7225 | 149 | |
AF179429.1 | - | 19 - 20 | 7332 | 149 |