The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286431.1 | NP_001273360.1 | 10 - 11 | 1551 | 77 |
NM_001286432.1 | NP_001273361.1 | 9 - 10 | 1444 | 77 | |
NM_002912.4 | NP_002903.3 | 8 - 9 | 1316 | 77 | |
XM_011536028.1 | XP_011534330.1 | 7 - 8 | 1407 | 77 | |
XM_011536029.2 | XP_011534331.1 | 7 - 8 | 1407 | 77 | |
XM_011536030.2 | XP_011534332.1 | 7 - 8 | 1407 | 77 | |
XM_011536031.2 | XP_011534333.1 | 8 - 9 | 1535 | 77 | |
XM_011536032.2 | XP_011534334.1 | 8 - 9 | 6125 | 77 | |
XM_011536036.2 | XP_011534338.1 | 7 - 8 | 1407 | 77 | |
XM_017011152.1 | XP_016866641.1 | 8 - 9 | 1535 | 77 | |
XM_017011153.1 | XP_016866642.1 | 8 - 9 | 1078 | 77 | |
XM_017011154.1 | XP_016866643.1 | 8 - 9 | 963 | 77 | |
XM_017011155.1 | XP_016866644.1 | 7 - 8 | 1407 | 77 | |
GenBank mRNA | AF035537.1 | - | 6 - 7 | 653 | 77 |
AF058701.1 | - | 8 - 9 | 943 | 77 | |
AF071798.1 | - | 7 - 8 | 1181 | 77 | |
AF078695.1 | - | 9 - 10 | 1446 | 77 | |
AF157476.1 | - | 7 - 8 | 889 | 77 | |
AF179428.1 | - | 8 - 9 | 1017 | 77 | |
AF179429.1 | - | 9 - 10 | 1124 | 77 | |
BC035823.1 | - | 7 - 8 | 1115 | 77 |