The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193465.1 | NP_001180394.1 | 8 - 9 | 2278 | 78 |
NM_001193466.1 | NP_001180395.1 | 7 - 8 | 2492 | 78 | |
NM_015443.3 | NP_056258.1 | 7 - 8 | 2194 | 78 | |
XM_006721823.1 | XP_006721886.1 | 8 - 9 | 2278 | 78 | |
XM_006721824.3 | XP_006721887.1 | 8 - 9 | 2462 | 78 | |
XM_011524628.2 | XP_011522930.1 | 8 - 9 | 2462 | 78 | |
XM_011524629.2 | XP_011522931.1 | 7 - 8 | 2360 | 78 | |
XM_011524630.2 | XP_011522932.1 | 8 - 9 | 2462 | 78 | |
XM_011524631.2 | XP_011522933.1 | 8 - 9 | 2463 | 78 | |
XM_011524632.2 | XP_011522934.1 | 7 - 8 | 888 | 78 | |
XM_017024488.1 | XP_016879977.1 | 8 - 9 | 2278 | 78 | |
XM_017024489.1 | XP_016879978.1 | 6 - 7 | 2287 | 78 | |
GenBank mRNA | AB033093.1 | - | 6 - 7 | 2115 | 78 |
AK291259.1 | - | 7 - 8 | 2255 | 78 | |
AL137317.1 | - | 3 - 4 | 436 | 78 | |
BC035892.1 | - | 2 - 3 | 299 | 78 | |
BC098376.1 | - | 7 - 8 | 2194 | 78 | |
BX538006.1 | - | 7 - 8 | 2495 | 78 |