The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017913.3 | NP_060383.2 | 2 - 3 | 621 | 151 |
GenBank mRNA | AK000497.1 | - | 2 - 3 | 616 | 151 |
AK000646.1 | - | 2 - 3 | 505 | 151 | |
AK225510.1 | - | 2 - 3 | 505 | 151 | |
BC014133.2 | - | 2 - 3 | 441 | 151 |