The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022481.5 | NP_071926.4 | 16 - 17 | 2426 | 107 |
XM_005268497.1 | XP_005268554.1 | 16 - 17 | 2390 | 107 | |
XM_005268498.1 | XP_005268555.1 | 16 - 17 | 2390 | 107 | |
XM_005268499.1 | XP_005268556.1 | 16 - 17 | 2364 | 107 | |
XM_005268500.3 | XP_005268557.1 | 16 - 17 | 2390 | 107 | |
XM_006714792.1 | XP_006714855.1 | 16 - 17 | 2390 | 107 | |
XM_011537677.2 | XP_011535979.1 | 14 - 15 | 1845 | 107 | |
GenBank mRNA | AJ310567.1 | - | 15 - 16 | 2376 | 107 |
AK091385.1 | - | 16 - 17 | 2304 | 107 | |
AK290052.1 | - | 16 - 17 | 2419 | 107 | |
AK295768.1 | - | 10 - 11 | 1403 | 107 |