The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022481.5 | NP_071926.4 | 27 - 28 | 3863 | 87 |
XM_005268497.1 | XP_005268554.1 | 27 - 28 | 3827 | 87 | |
XM_005268498.1 | XP_005268555.1 | 27 - 28 | 3767 | 87 | |
XM_005268499.1 | XP_005268556.1 | 27 - 28 | 3801 | 87 | |
XM_006714792.1 | XP_006714855.1 | 26 - 27 | 3614 | 87 | |
XM_011537677.2 | XP_011535979.1 | 25 - 26 | 3282 | 87 | |
GenBank mRNA | AJ310567.1 | - | 26 - 27 | 3813 | 87 |
AK001579.1 | - | 10 - 11 | 1787 | 87 | |
AK091385.1 | - | 26 - 27 | 3529 | 87 | |
AK290052.1 | - | 27 - 28 | 3856 | 87 | |
AK295768.1 | - | 21 - 22 | 2840 | 87 | |
BC009968.2 | - | 1 - 2 | 114 | 87 |