The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257291.1 | NP_001244220.1 | 9 - 10 | 1381 | 69 |
NM_032591.2 | NP_115980.1 | 9 - 10 | 1378 | 69 | |
XM_005272681.4 | XP_005272738.1 | 9 - 10 | 1371 | 69 | |
XM_005272682.4 | XP_005272739.1 | 9 - 10 | 1371 | 69 | |
XM_006724564.3 | XP_006724627.1 | 9 - 10 | 1371 | 69 | |
XM_011543990.2 | XP_011542292.1 | 9 - 10 | 1371 | 69 | |
XM_017029905.1 | XP_016885394.1 | 9 - 10 | 1371 | 69 | |
XM_017029906.1 | XP_016885395.1 | 9 - 10 | 1371 | 69 | |
GenBank mRNA | AF298591.1 | - | 9 - 10 | 1243 | 69 |
AK056599.1 | - | 7 - 8 | 706 | 69 |