Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSPP1
Entrez Gene ID
79848
Gene Name
centrosome and spindle pole associated protein 1
Gene Aliases
CSPP, JBTS21
Chromosome Location
Chr.8: 67062417 - 67196614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291339.1 NP_001278268.1 24 - 25 3350 73
NM_024790.6 NP_079066.5 27 - 28 3350 73
XM_005251305.3 XP_005251362.2 30 - 31 3834 73
XM_006716474.2 XP_006716537.2 29 - 30 3681 73
XM_006716477.2 XP_006716540.2 25 - 26 3303 73
XM_011517598.1 XP_011515900.1 31 - 32 3876 73
XM_011517599.1 XP_011515901.1 30 - 31 3852 73
XM_011517600.1 XP_011515902.1 29 - 30 3810 73
XM_011517601.1 XP_011515903.1 29 - 30 3771 73
XM_011517602.1 XP_011515904.1 28 - 29 3729 73
XM_011517603.1 XP_011515905.1 30 - 31 3559 73
XM_011517607.1 XP_011515909.1 29 - 30 3439 73
XM_011517608.1 XP_011515910.1 28 - 29 3436 73
XM_011517609.1 XP_011515911.1 27 - 28 3558 73
XM_011517611.2 XP_011515913.1 15 - 16 1935 73
XM_017013847.1 XP_016869336.1 29 - 30 3735 73
XM_017013848.1 XP_016869337.1 28 - 29 3711 73
XM_017013849.1 XP_016869338.1 28 - 29 3672 73
XM_017013850.1 XP_016869339.1 27 - 28 3630 73
XM_017013851.1 XP_016869340.1 27 - 28 3394 73
XM_017013852.1 XP_016869341.1 26 - 27 3477 73
XM_017013853.1 XP_016869342.1 26 - 27 3241 73
XM_017013854.1 XP_016869343.1 24 - 25 3279 73
XM_017013855.1 XP_016869344.1 22 - 23 3045 73
XM_017013856.1 XP_016869345.1 22 - 23 2863 73
XM_017013857.1 XP_016869346.1 23 - 24 2913 73
XM_017013858.1 XP_016869347.1 21 - 22 2661 73
XM_017013859.1 XP_016869348.1 17 - 18 2063 73
GenBank mRNA AJ583433.1 - 24 - 25 3350 73
AK026143.1 - - 797 73
AM156947.1 - 27 - 28 3351 73
BC022867.1 - 9 - 10 849 73
BX649147.1 - 27 - 28 3352 73

View Full Product Details