The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199207.1 | NP_001186136.1 | 6 - 7 | 1406 | 79 |
NM_005685.3 | NP_005676.3 | 6 - 7 | 1310 | 79 | |
NM_016328.2 | NP_057412.1 | 6 - 7 | 1310 | 79 | |
XM_006716182.3 | XP_006716245.1 | 6 - 7 | 1106 | 79 | |
XM_006716183.3 | XP_006716246.1 | 6 - 7 | 951 | 79 | |
XM_011516713.1 | XP_011515015.1 | 6 - 7 | 1106 | 79 | |
XM_017012802.1 | XP_016868291.1 | 6 - 7 | 1106 | 79 | |
XM_017012803.1 | XP_016868292.1 | 6 - 7 | 1106 | 79 | |
XM_017012804.1 | XP_016868293.1 | 6 - 7 | 1035 | 79 | |
XM_017012805.1 | XP_016868294.1 | 6 - 7 | 1106 | 79 | |
GenBank mRNA | AB209389.1 | - | 6 - 7 | 2608 | 79 |
AF089107.1 | - | 5 - 6 | 924 | 79 | |
AF104923.1 | - | 6 - 7 | 956 | 79 | |
AF118270.2 | - | 6 - 7 | 965 | 79 | |
AF151354.1 | - | 6 - 7 | 1310 | 79 | |
AF156489.1 | - | 6 - 7 | 1013 | 79 | |
AY648295.1 | - | 6 - 7 | 1226 | 79 | |
BC018136.1 | - | 6 - 7 | 1075 | 79 |