The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005498.3 | NP_001005498.2 | 2 - 3 | 372 | 63 |
XM_005257670.1 | XP_005257727.1 | 3 - 4 | 404 | 63 | |
XM_011525249.2 | XP_011523551.1 | 3 - 4 | 1851 | 63 | |
XM_011525250.2 | XP_011523552.1 | 4 - 5 | 561 | 63 | |
XM_011525251.2 | XP_011523553.1 | 3 - 4 | 378 | 63 | |
XM_017025079.1 | XP_016880568.1 | 3 - 4 | 808 | 63 | |
GenBank mRNA | AK057882.1 | - | 1 - 2 | 156 | 63 |
AK074034.1 | - | 2 - 3 | 384 | 63 | |
AK292166.1 | - | 2 - 3 | 327 | 63 | |
AK303434.1 | - | 1 - 2 | 188 | 63 | |
AY500247.1 | - | 2 - 3 | 223 | 63 | |
BP284970.1 | - | 3 - 4 | 446 | 63 | |
BP288209.1 | - | 3 - 4 | 446 | 63 | |
DC337840.1 | - | 3 - 4 | 447 | 63 | |
DC348660.1 | - | 1 - 2 | 124 | 63 |