The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022842.4 | NP_073753.3 | 5 - 6 | 1380 | 73 |
XM_011534024.1 | XP_011532326.1 | 5 - 6 | 1338 | 73 | |
XM_017007070.1 | XP_016862559.1 | 5 - 6 | 1366 | 73 | |
GenBank mRNA | AF468010.1 | - | 5 - 6 | 1355 | 73 |
AK023834.1 | - | 3 - 4 | 958 | 73 | |
AK026187.1 | - | 2 - 3 | 397 | 73 | |
AK026622.1 | - | 5 - 6 | 1355 | 73 | |
AK314441.1 | - | 5 - 6 | 1344 | 73 | |
AY026461.1 | - | 5 - 6 | 1327 | 73 | |
AY167484.1 | - | 5 - 6 | 1337 | 73 | |
AY375452.1 | - | 5 - 6 | 1326 | 73 | |
BC069254.1 | - | 5 - 6 | 1333 | 73 | |
KU215672.1 | - | 4 - 5 | 1158 | 73 | |
KU257676.1 | - | 4 - 5 | 1155 | 73 |