The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022842.4 | NP_073753.3 | 8 - 9 | 2211 | 106 |
XM_011534024.1 | XP_011532326.1 | 8 - 9 | 2169 | 106 | |
GenBank mRNA | AF468010.1 | - | 8 - 9 | 2186 | 106 |
AK023834.1 | - | 6 - 7 | 1789 | 106 | |
AK026622.1 | - | 8 - 9 | 2186 | 106 | |
AK314441.1 | - | 8 - 9 | 2175 | 106 | |
AY026461.1 | - | 8 - 9 | 2158 | 106 | |
AY167484.1 | - | 8 - 9 | 2168 | 106 |