The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144683.3 | NP_653284.2 | 2 - 3 | 371 | 65 |
GenBank mRNA | AY358566.1 | - | 2 - 3 | 371 | 65 |
BC015582.2 | - | 1 - 2 | 361 | 65 |