The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013397.5 | NP_037529.3 | 5 - 6 | 617 | 89 |
GenBank mRNA | BC110459.1 | - | 3 - 4 | 1069 | 89 |
DA917216.1 | - | 1 - 2 | 231 | 89 |