Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBR2
Entrez Gene ID
23304
Gene Name
ubiquitin protein ligase E3 component n-recognin 2
Gene Aliases
C6orf133, bA49A4.1, dJ242G1.1, dJ392M17.3
Chromosome Location
Chr.6: 42563905 - 42693505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015255.2 NP_056070.1 37 - 38 4486 74
XM_005248965.4 XP_005249022.1 37 - 38 4603 74
XM_005248966.3 XP_005249023.1 36 - 37 3886 74
XM_011514438.2 XP_011512740.2 37 - 38 4686 74
XM_011514439.1 XP_011512741.1 28 - 29 3053 74
XM_011514440.1 XP_011512742.1 28 - 29 3053 74
XM_011514441.2 XP_011512743.1 25 - 26 2666 74
XM_017010594.1 XP_016866083.1 37 - 38 4687 74
XM_017010595.1 XP_016866084.1 37 - 38 4684 74
XM_017010596.1 XP_016866085.1 37 - 38 4518 74
GenBank mRNA AB002347.1 - 25 - 26 2748 74
AK026998.1 - 16 - 17 1749 74
AK125795.1 - 29 - 30 3163 74
AK127310.1 - 1 - 2 656 74
AK304070.1 - 29 - 30 3139 74
AY061884.1 - 37 - 38 4188 74
BX647467.1 - 19 - 20 3142 74

View Full Product Details