Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COMMD4
Entrez Gene ID
54939
Gene Name
COMM domain containing 4
Gene Aliases
-
Chromosome Location
Chr.15: 75335996 - 75341927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284377.1 NP_001271306.1 2 - 3 164 58
NM_001284378.1 NP_001271307.1 2 - 3 164 58
NM_001321844.1 NP_001308773.1 2 - 3 158 58
NM_017828.4 NP_060298.2 2 - 3 164 58
NR_104312.1 2 - 3 164 58
NR_104313.1 2 - 3 164 58
XM_005254509.3 XP_005254566.1 2 - 3 190 58
XM_011521739.1 XP_011520041.1 2 - 3 199 58
XM_011521741.1 XP_011520043.1 2 - 3 199 58
GenBank mRNA AK000459.1 - 2 - 3 101 58
AK225716.1 - 2 - 3 125 58
AK293708.1 - 2 - 3 100 58
AK314740.1 - 2 - 3 97 58
AL136872.1 - 2 - 3 84 58
AY542160.1 - 1 - 2 70 58
BC000837.2 - 1 - 2 67 58
BC068998.1 - 2 - 3 86 58
BC069195.1 - 2 - 3 92 58
BI829264.1 - 2 - 3 140 58
BI829484.1 - 2 - 3 164 58
BU540913.1 - 2 - 3 106 58
CR533452.1 - 1 - 2 70 58

View Full Product Details