Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COMMD4
Entrez Gene ID
54939
Gene Name
COMM domain containing 4
Gene Aliases
-
Chromosome Location
Chr.15: 75335996 - 75341927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284377.1 NP_001271306.1 6 - 7 477 59
NM_001284379.1 NP_001271308.1 5 - 6 455 59
NM_001321846.1 NP_001308775.1 5 - 6 411 59
XM_011521741.1 XP_011520043.1 6 - 7 512 59
XM_017022385.1 XP_016877874.1 5 - 6 446 59
GenBank mRNA AK000459.1 - 6 - 7 414 59
AW117823.1 - 1 - 2 59
BC000837.2 - 5 - 6 380 59
BC069195.1 - 6 - 7 352 59
BQ233598.1 - - 440 59

View Full Product Details