The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000823.3 | NP_000814.2 | 3 - 4 | 315 | 67 |
GenBank mRNA | AY557192.1 | - | 3 - 4 | 267 | 67 |
L01406.1 | - | 3 - 4 | 318 | 67 | |
L09237.1 | - | 3 - 4 | 315 | 67 |