The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000823.3 | NP_000814.2 | 7 - 8 | 795 | 60 |
XM_011515263.1 | XP_011513565.1 | 4 - 5 | 813 | 60 | |
GenBank mRNA | AB058895.1 | - | 2 - 3 | 323 | 60 |
AF282259.1 | - | 4 - 5 | 813 | 60 | |
AY557192.1 | - | 7 - 8 | 747 | 60 | |
L01406.1 | - | 7 - 8 | 798 | 60 | |
L09237.1 | - | 7 - 8 | 795 | 60 |