The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052897.3 | NP_443129.3 | 3 - 4 | 341 | 107 |
XM_005268617.1 | XP_005268674.1 | 3 - 4 | 394 | 107 | |
XM_005268618.1 | XP_005268675.1 | 3 - 4 | 490 | 107 | |
XM_005268620.2 | XP_005268677.1 | 3 - 4 | 394 | 107 | |
XM_006719217.1 | XP_006719280.1 | 3 - 4 | 239 | 107 | |
XM_006719218.1 | XP_006719281.1 | 3 - 4 | 227 | 107 | |
XM_011537851.2 | XP_011536153.1 | 3 - 4 | 1227 | 107 | |
GenBank mRNA | AK056399.1 | - | 3 - 4 | 375 | 107 |
AK296990.1 | - | 3 - 4 | 375 | 107 | |
BC065530.1 | - | 3 - 4 | 341 | 107 |