The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022916.4 | NP_075067.2 | 6 - 7 | 887 | 98 |
GenBank mRNA | AF439857.1 | - | 6 - 7 | 798 | 98 |
AK026840.1 | - | 6 - 7 | 797 | 98 | |
AK315545.1 | - | 6 - 7 | 825 | 98 | |
BC016617.1 | - | 6 - 7 | 795 | 98 |